Sodium isopropoxide

Supplier: Thermo Scientific Chemicals
Danger

89446-06 89446-14 89446-22
CAAA89446-06EA 85.64 CAD
CAAA89446-06 CAAA89446-14 CAAA89446-22
Sodium isopropoxide
Sodium isopropoxide

CAS No.: 683-60-3 Molecular Formula: C3H7NaO Formula Weight: 82.08 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 70-75° UN No.: 3206 MDL No.: MFCD00061566

Formula: C₃H₇NaO
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00061566
CAS Number: 683-60-3
EINECS: 211-673-7
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR