Silica gel ≥99.5%, large pore

Supplier: Thermo Scientific Chemicals
Warning

89385-36 89385-A3
CAAA89385-36EA 145.04 CAD
CAAA89385-36 CAAA89385-A3
Silica gel ≥99.5%, large pore
Silica gel

Purity: 99.5% CAS No.: 63231-67-4 Physical Form: 56 Micron, S.A. 500m2/g, P.V. 0.75cc/g Appearance: White MDL No.: MFCD00011232

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR