Aluminum di(isopropoxide)acetoacetic ester chelate (≥9,6% Al)
Supplier: Thermo Scientific Chemicals
|
Warning
|
89350-22
89350-36
CAAA89350-22EA
105.7
CAD
CAAA89350-22
CAAA89350-36
Aluminum di(isopropoxide)acetoacetic ester chelate (≥9,6% Al)
Aluminum di(isopropoxide)acetoacetic ester chelate
Purity: 9.6% min
CAS No.: 14782-75-3
Molecular Formula: C12H23AlO5
Formula Weight: 274.3
Flash Point: 50°(122°F)
Physical Form: Liquid
Density: 1.05
Sensitivity: Moisture Sensitive
Odor: Alcohol-like
UN No.: 1993
MDL No.: MFCD00135521
Formula:
C₁₂H₂₃AlO₅ Density: 1,05 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00135521 CAS Number: 14782-75-3 EINECS: 238-851-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...