Gadolinium(III) acetate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
89158-06
89158-14
89158-22
CAAA89158-06EA
45.19
CAD
CAAA89158-06
CAAA89158-14
CAAA89158-22
Gadolinium(III) acetate hydrate ≥99.9%
Gadolinium(III) acetate hydrate
Soluble in water
Purity: 99.9%
CAS No.: 100587-93-7
Molecular Formula: Gd(OOCCH3)3.xH2O
Formula Weight: 334.39(anhy)
Physical Form: Crystalline
Density: 1.611
Sensitivity: Hygroscopic
Odor: Odorless
Boiling Point: dec.
Solubility: Soluble in water
MDL No.: MFCD00150116
Formula:
Gd(OOCCH₃)₃·xH₂O Boiling Pt: Dec. Density: 1,611 Storage Temperature: Ambient |
MDL Number:
MFCD00150116 CAS Number: 100587-93-7 EINECS: 240-204-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...