Borane dimethylamine complex ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
89149-14
89149-22
CAAA89149-14EA
128.93
CAD
CAAA89149-14
CAAA89149-22
Borane dimethylamine complex ≥97%
Borane dimethylamine complex
Purity: 97%
CAS No.: 74-94-2
Molecular Formula: BH3.NH(CH3)2
Formula Weight: 58.92
Physical Form: Crystalline
Density: 0.69
Appearance: White
Melting Point: 36°
Boiling Point: 59-65°/1-2mm
UN No.: 2925
MDL No.: MFCD00051068
Formula:
C₂H₁₀BN Boiling Pt: 59…65 °C (1…2 mmHg) Melting Pt: 36 °C Density: 0,69 Storage Temperature: Ambient |
MDL Number:
MFCD00051068 CAS Number: 74-94-2 EINECS: 200-823-7 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...