Amberlyst® 15 (H), Amberlyst™ ion exchange resin
Supplier: Thermo Scientific Chemicals
89079-18
89079-30
89079-A1
CAAA89079-18EA
47.72
CAD
CAAA89079-18
CAAA89079-30
CAAA89079-A1
Amberlyst® 15 (H), Amberlyst™ ion exchange resin
Amberlyst® 15 (H)
Fieser: 5,355 6,302 9,256 11,276 13,152 15,178 18,194
CAS No.: 39389-20-3
Molecular Formula: Styrene-DVB; Strongly acidic macroreticular resin
Physical Form: Beads
Appearance: Pale brown
Odor: Odorless
MDL No.: MFCD00145841
Formula:
(C₁₀H₁₀·C₈H₈O₃S)x Storage Temperature: Ambient |
MDL Number:
MFCD00145841 CAS Number: 39389-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...