Magnesium hexafluorosilicate hydrate ≥99%

Supplier: Thermo Scientific Chemicals
Danger

89018-36 89018-A3
CAAA89018-36EA 111.71 CAD
CAAA89018-36 CAAA89018-A3
Magnesium hexafluorosilicate hydrate ≥99%
Magnesium hexafluorosilicate hydrate

Molecular Formula: MgSiF6.xH2O Formula Weight: 166.39(anhy) Physical Form: Powder Density: 1.788 Melting Point: 120° dec. UN No.: 2853 MDL No.: MFCD00016196

Formula: F₆MgSi·H₂O
Melting Pt: 120 °C (dec.)
Density: 1.788
Storage Temperature: Ambient
MDL Number: MFCD00016196
CAS Number: 16949-65-8
EINECS: 241-022-2
UN: 2853
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR