Tetraammineplatinum(II) nitrate

Supplier: Thermo Scientific Chemicals
Danger

88960-03 88960-06
CAAA88960-03EA 298.62 CAD
CAAA88960-03 CAAA88960-06
Tetraammineplatinum(II) nitrate
Tetraammineplatinum(II) nitrate

CAS No.: 20634-12-2 Molecular Formula: H12N6O6Pt Formula Weight: 387.21 Physical Form: Powder/chunks Appearance: White Melting Point: 262° dec. UN No.: 1477 MDL No.: MFCD00011624

Formula: H₁₂N₆O₆Pt
Melting Pt: 262 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR