Lithium trifluoroacetate monohydrate ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
88938-09
88938-18
CAAA88938-09EA
68.57
CAD
CAAA88938-09
CAAA88938-18
Lithium trifluoroacetate monohydrate ≥97%
Lithium trifluoroacetate monohydrate
Purity: 97%
CAS No.: 2923-17-3
Molecular Formula: C2F3LiO2.H2O
Formula Weight: 137.97 (119.96anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
UN No.: 2811
MDL No.: MFCD00149663
Formula:
C₂F₃LiO₂·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149663 CAS Number: 2923-17-3 EINECS: 220-878-0 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...