Iron(II) gluconate hydrate ≥97%
Supplier: Thermo Scientific Chemicals
88726-36
88726-A3
CAAA88726-36EA
91.01
CAD
CAAA88726-36
CAAA88726-A3
Iron(II) gluconate hydrate ≥97%
Iron(II) gluconate hydrate
Purity: 97%
CAS No.: 22830-45-1
Molecular Formula: Fe[HOCH2(CHOH)4CO2]2.xH2O (x≈2)
Formula Weight: 446.19(anhy)
Physical Form: Granular
Melting Point: ca 140°
MDL No.: MFCD00150872
Formula:
C₁₂H₂₄FeO₁₅ Melting Pt: ∼ 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150872 CAS Number: 22830-45-1 EINECS: 206-076-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...