Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1 L, Thermo Scientific

Supplier: Thermo Scientific Chemicals

88632-LQ 88632-LS
CAAA88632-LQEA 121.98 CAD
CAAA88632-LQ CAAA88632-LS
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1 L, Thermo Scientific
Standards Environmental Analysis Standards

One unit makes one liter of 1% solution after dilution


CAS No.: 9005-84-9
Physical Form: Liquid, For iodometric titrations
MDL No.: MFCD00082026

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR