Ammonia solution concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
88576-LQ
88576-LS
CAAA88576-LQEA
81.69
CAD
CAAA88576-LQ
CAAA88576-LS
Ammonia solution concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Ammonia solution
One unit makes one liter of 0.1N solution after dilution
- Physical Form: Liquid
- Appearance: Colorless
- Odor: Ammonia-like
Formula:
NH₃ Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...