Iron nickel oxide, tech.

Supplier: Thermo Scientific Chemicals
Danger

88530-09 88530-18
CAAA88530-09EA 94.17 CAD
CAAA88530-09 CAAA88530-18
Iron nickel oxide, tech.
Iron nickel oxide

CAS No.: 12168-54-6 Molecular Formula: NiFe2O4 Formula Weight: 234.39 Physical Form: Powder Density: 5.368 Odor: Odorless MDL No.: MFCD00016254

Formula: NiFe₂O₄
Density: 5,368
Storage Temperature: Ambient
MDL Number: MFCD00016254
CAS Number: 12168-54-6
EINECS: 235-335-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR