Titanium aluminide ≥99.5% (metals basis) -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
88393-09
88393-18
CAAA88393-09EA
106.49
CAD
CAAA88393-09
CAAA88393-18
Titanium aluminide ≥99.5% (metals basis) -325 mesh
Titanium aluminide
Purity: 99.5%
CAS No.: 12003-96-2
Molecular Formula: TiAl
Sensitivity: Air Sensitive
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00210656
Formula:
AlTi Storage Temperature: Ambient |
MDL Number:
MFCD00210656 CAS Number: 12003-96-2 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...