Zinc nitride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

88212-04 88212-09
CAAA88212-04EA 128.77 CAD
CAAA88212-04 CAAA88212-09
Zinc nitride ≥99% (metals basis)
Zinc nitride

Purity: 99% CAS No.: 1313-49-1 Molecular Formula: Zn3N2 Formula Weight: 224.12 Physical Form: Powder Density: 6.22 MDL No.: MFCD00061327

Formula: N₂Zn₃
Density: 6,22
Storage Temperature: Ambient
MDL Number: MFCD00061327
CAS Number: 1313-49-1
EINECS: 215-207-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR