Iron nitride
Supplier: Thermo Scientific Chemicals
88198-14
88198-22
CAAA88198-14EA
339.7
CAD
CAAA88198-14
CAAA88198-22
Iron nitride
Iron nitride
CAS No.: 37245-77-5
Molecular Formula: FexN (x=2-4)
Physical Form: -325 Mesh Powder
MDL No.: MFCD00134717
Formula:
Fe_x N (x=2-4) Storage Temperature: Ambient |
MDL Number:
MFCD00134717 CAS Number: 37245-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...