Iron boride 98%

Supplier: Thermo Scientific Chemicals

88146-14 88146-22
CAAA88146-14EA 270.18 CAD
CAAA88146-14 CAAA88146-22
Iron boride 98%
Iron boride

Purity: 98% CAS No.: 12006-84-7 Molecular Formula: FeB Formula Weight: 66.66 Physical Form: Powder Density: 7.15 MDL No.: MFCD00016092

Formula: BFe
Density: 7,15
Storage Temperature: Ambient
MDL Number: MFCD00016092
CAS Number: 12006-84-7
EINECS: 234-489-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR