Diphenylzinc 98+%

Supplier: Thermo Scientific Chemicals
Warning

87908-03 87908-06
CAAA87908-03EA 230.68 CAD
CAAA87908-03 CAAA87908-06
Diphenylzinc 98+%
Diphenylzinc

Purity: 98+% CAS No.: 1078-58-6 Molecular Formula: (C6H5)2Zn Formula Weight: 219.58 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Boiling Point: 280-285° UN No.: 3181 MDL No.: MFCD00014071

Formula: C₁₂H₁₀Zn
Boiling Pt: 280…285 °C
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00014071
CAS Number: 1078-58-6
EINECS: 214-082-2
UN: 3181
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR