Reinecke's salt monohydrate ≥93.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium reineckate, Ammonium diamminetetrathiocyanatochromate(III) monohydrate

87777-14 87777-22
CAAA87777-14EA 114.55 CAD
CAAA87777-14 CAAA87777-22
Reinecke's salt monohydrate ≥93.0% ACS
Reinecke's salt monohydrate

Used to precipitate primary and secondary amines as their ammonium salts
Insoluble in dilute hydrochloric acid. Moderately soluble in water. Soluble in alcohol


Purity: 93.0% min CAS No.: 13573-16-5 Molecular Formula: C4H10CrN7S4.H2O Formula Weight: 354.44 (336.42anhy) Physical Form: Powder Appearance: Dark red Odor: Odorless Solubility: Insoluble in dilute hydrochloric acid. Moderately soluble in water. Soluble in alcohol Merck Reference: 14,8128 UN No.: 3077 MDL No.: MFCD00066638


Used to precipitate primary and secondary amines as their ammonium salts

Formula: C₄H₁₀CrN₇S₄·1H₂O
MW: 354.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066638
CAS Number: 13573-17-6
EINECS: 237-003-3
UN: 3077
ADR: 9,III
Merck Index: 14,8128

Order Now

Specification Test Results

Insoluble in dilute hydrochloric acid 0.05% max.
Sensitivity Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR