Sodium borohydride 4.4 M in aqueous solution stable
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
87659-AE
87659-AP
87659-K3
CAAA87659-AEEA
51.67
CAD
CAAA87659-AE
CAAA87659-AP
CAAA87659-K3
Sodium borohydride 4.4 M in aqueous solution stable
Sodium borohydride
CAS No.: 16940-66-2
Molecular Formula: BH4Na
Formula Weight: 37.83
Physical Form: Liquid
Density: 1.4
Sensitivity: Air Sensitive
Appearance: Colorless
UN No.: 3320
MDL No.: MFCD00003518
Formula:
NaBH₄ MW: 37.83 g/mol Melting Pt: 400 Density: 1.4 Flash Pt: 69 Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 3320 ADR: 8 II Merck Index: 13,08664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...