Zinc tetrafluoroborate hydrate (≥18% Zn)

Supplier: Thermo Scientific Chemicals
Danger

85124-09 85124-18 85124-30
CAAA85124-09EA 38.71 CAD
CAAA85124-09 CAAA85124-18 CAAA85124-30
Zinc tetrafluoroborate hydrate (≥18% Zn)
Zinc tetrafluoroborate hydrate

CAS No.: 27860-83-9 Molecular Formula: Zn(BF4)2.xH2O Formula Weight: 238.98(anhy) Physical Form: Powder Sensitivity: Hygroscopic Odor: Odorless UN No.: 3260 MDL No.: MFCD00150371

Formula: Zn(BF₄)₂·xH₂O
MW: 238.98 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150371
CAS Number: 27860-83-9
EINECS: 237-534-0
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR