Dibutyltin bis(2-ethylhexanoate)
Supplier: Thermo Scientific Chemicals
|
Danger
|
71126-06
71126-14
71126-22
CAAA71126-06EA
62.73
CAD
CAAA71126-06
CAAA71126-14
CAAA71126-22
Dibutyltin bis(2-ethylhexanoate)
Dibutyltin bis(2-ethylhexanoate)
Soluble in mineral oil, acetone, xylene
CAS No.: 2781-10-4
Molecular Formula: C24H48O4Sn
Formula Weight: 519.34
Physical Form: Lump
Density: 1.070
Odor: Odorless
Boiling Point: 220°/2mm
Solubility: Soluble in mineral oil, acetone, xylene
UN No.: 3146
MDL No.: MFCD00015261
Formula:
C₂₄H₄₈O₄Sn Boiling Pt: 220 °C (2 mmHg) Melting Pt: 57…59 °C Density: 1.070 Storage Temperature: Ambient |
MDL Number:
MFCD00015261 CAS Number: 2781-10-4 EINECS: 220-481-2 UN: 3146 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...