Potassium selenocyanate ≥98.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
68107-18
68107-09
CAAA-68107-18EA
508.76
CAD
CAAA-68107-18
CAAA68107-09
Potassium selenocyanate ≥98.5%
Potassium selenocyanate
Purity: 98.5%
CAS No.: 3425-46-5
Molecular Formula: KSeCN
Formula Weight: 144.08
Physical Form: Crystalline
Density: 2.347
Appearance: White to off-white
Odor: Odorless
Melting Point: 162-164°
UN No.: 3283
MDL No.: MFCD00011366
Formula:
CKNSe Melting Pt: 162…164 °C Density: 2.347 Storage Temperature: Ambient |
MDL Number:
MFCD00011366 CAS Number: 3425-46-5 EINECS: 222-320-1 UN: 3283 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...