Silicon nitride α-phase ≥99.85% (metals basis) 90+% α-phase, Electronic Grade
Supplier: Thermo Scientific Chemicals
49113-09
49113-18
CAAA49113-09EA
42.5
CAD
CAAA49113-09
CAAA49113-18
Silicon nitride α-phase ≥99.85% (metals basis) 90+% α-phase, Electronic Grade
Silicon nitride
Purity: 99.85%
CAS No.: 12033-89-5
Molecular Formula: N4Si3
Formula Weight: 140.28
Physical Form: -325 Mesh Powder
Density: 3.2
Appearance: Pale gray
Odor: Odorless
Melting Point: 1900° subl.
Merck Reference: 14,8497
MDL No.: MFCD00011230
Formula:
N₄Si₃ Melting Pt: 1900 °C (subl.) Density: 3.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011230 CAS Number: 12033-89-5 EINECS: 234-796-8 Merck Index: 14,8497 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...