Bis(tricyclohexylphosphine)palladium(0) ≥97%

Supplier: Thermo Scientific Chemicals
Warning

45792-03 45792-MD
CAAA45792-03EA 1017.52 CAD
CAAA45792-03 CAAA45792-MD
Bis(tricyclohexylphosphine)palladium(0) ≥97%
Bis(tricyclohexylphosphine)palladium(0)

Purity: 97% min CAS No.: 33309-88-5 Molecular Formula: C36H66P2Pd Formula Weight: 667.29 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: Off-white Odor: Odorless MDL No.: MFCD01073796

Formula: C₃₆H₆₆P₂Pd
MW: 667.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01073796
CAS Number: 33309-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR