Carbon black ≥99.9%, acetylene, 100% compressed
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Furnace black, Lamp Black
45527-30
45527-95
AA45527-30EA
102.86
CAD
AA45527-30
CAAA45527-95
Carbon black ≥99.9%, acetylene, 100% compressed
Carbon black
Purity: 99.9+%
CAS No.: 1333-86-4
Physical Form: S.A. 75m2/g, Bulk density 170-230g/L
Appearance: Black
Odor: Odorless
UN No.: 1325
MDL No.: MFCD00133992
Formula:
C MW: 12.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 1333-86-4 EINECS: 215-609-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...