Toray Carbon Paper, Thermo Scientific
Supplier: Thermo Scientific Chemicals
045365-KS 45356-KS
AA45365-KSEA 290.26 CAD
AA45365-KS CAAA45356-KS
Toray Carbon Paper, Thermo Scientific
Paper
Toray carbon paper, TGP-H-60 can be used as a gas diffusion layer in fuel cell applications. It is also suitable for use as a catalyst backing layer.
  • Insoluble in water
  • Air and light sensitive
  • Container must be closed tightly and stored in a cool, dry and well-ventilated place
It is incompatible with strong oxidizing agents, chlorinated paraffins, lead oxide, manganese oxide, iron oxide, liquid oxygen, oils, chlorinetrifluoride, fluoride and potassium superoxide.
Order Now
SPECIFICATIONS
Environmentally Preferable