Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform (20,6% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct, Tris(dibenylideneacetone)dipalladium-chloroform, Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Fieser: 11,508 13,9 14,339 15,353 16,372 17,394 19,390 20,420 21,474
CAS No.: 52522-40-4
Molecular Formula: C51H42O3Pd2.CHCl3
Formula Weight: 1035.11
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Melting Point: 131-135°
MDL No.: MFCD00075479
Formula:
C₅₁H₄₂O₃Pd₂·CHCl₃ Melting Pt: 131…135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075479 CAS Number: 52522-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...