Zinc oxide 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, NanoArc® ZN-0625
Supplier: Thermo Scientific Chemicals
|
Warning
|
44924-22
44924-A3
CAAA44924-22EA
58.46
CAD
CAAA44924-22
CAAA44924-A3
Zinc oxide 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, NanoArc® ZN-0625
Zinc oxide
Zinc oxide dispersion in Dowanol® PMA suitable for incorporation in a wide variety of coatings formulations
Sold in collaboration with Nanophase Technologies Corporation
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: 70nm APS for dry powder (typical)
Odor: Ester-like
UN No.: 1993
MDL No.: MFCD00011300
Applications: Zinc oxide dispersion in Dowanol® PMA suitable for incorporation in a wide variety of coatings formulations
Formula:
ZnO Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...