(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised

Supplier: Thermo Scientific Chemicals


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 50 ppm-100 ppm

44793-06 44793-14
CAAAA44793-06EA 153.1 CAD
CAAAA44793-06 CAAAA44793-14
(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised
(Perfluorocyclohexyl)methyl acrylate

Stab. with 50ppm to 100ppm 4-methoxyphenol


Purity: 96% CAS No.: 40677-94-9 Molecular Formula: C10H5F11O2 Formula Weight: 366.13 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Air Sensitive Appearance: Colorless MDL No.: MFCD00080614

Formula: C₁₀H₅F₁₁O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00080614
CAS Number: 40677-94-9

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR