Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Palladium(I) tri-tert-butylphosphine bromide dimer, Di-mu-bromobis(tri-t-butylphosphino)dipalladium(I)

44446-02 44446-04 44446-77
CAAA44446-02EA 480.32 CAD
CAAA44446-02 CAAA44446-04 CAAA44446-77
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)

Applications: Coupling reactions. Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides. Especially active in difficult aminations


CAS No.: 185812-86-6 Molecular Formula: C24H54Br2P2Pd2 Formula Weight: 777.29 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Air Sensitive Appearance: Dark green Solubility: Soluble in benzene and toluene MDL No.: MFCD04114019


Applications: Coupling reactions. Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides. Especially active in difficult aminations

Formula: C₂₄H₅₆Br₂P₂Pd₂
MW: 779.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04114019
CAS Number: 185812-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR