Alkali Metals, plasma standard solution, Specpure®, Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr @ 100µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
44428-AE
44428-AP
CAAA44428-AEEA
306.52
CAD
CAAA44428-AE
CAAA44428-AP
Alkali Metals, plasma standard solution, Specpure®, Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr @ 100µg/ml, Thermo Scientific
Standards
Cationic Standards
Multi-Element Standards
Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD08061357
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...