Silica gel, granules -6+16 mesh desiccant, with indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
44389-30
44389-A1
44389-A7
CAAA44389-30EA
65.89
CAD
CAAA44389-30
CAAA44389-A1
CAAA44389-A7
Silica gel, granules -6+16 mesh desiccant, with indicator
Silica gel
Re-use after being saturated with water by heating to 121 °C for approximately 8 hours
CAS No.: 7631-86-9
Physical Form: Granules
Sensitivity: Hygroscopic
Appearance: Blue to red
Odor: Odorless
pH: 6
MDL No.: MFCD00011232
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...