Ruthenium 5% on activated carbon wetted with ca. 50% water, powder, reduced

Supplier: Thermo Scientific Chemicals

44338-06 44338-14 44338-22
CAAA44338-06EA 77.42 CAD
CAAA44338-06 CAAA44338-14 CAAA44338-22
Ruthenium 5% on activated carbon wetted with ca. 50% water, powder, reduced
Ruthenium

Sold on dry weight basis. Unit weight excludes water weight


CAS No.: 7440-18-8 Odor: Odorless MDL No.: MFCD03458417

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD03458417
CAS Number: 7440-18-8
EINECS: 231-127-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR