Palladium 5% on activated carbon paste (50% water) 5R39
Supplier: Thermo Scientific Chemicals
44337-06
44337-14
44337-22
CAAA44337-06EA
130.67
CAD
CAAA44337-06
CAAA44337-14
CAAA44337-22
Palladium 5% on activated carbon paste (50% water) 5R39
Palladium
Applications: Debenzylation, C-N and C-O cleavage, alkene hydrogenation
Sold on a dry weight basis. Unit weight excludes water weight
CAS No.: 7440-05-3
Appearance: Black
Odor: Odorless
MDL No.: MFCD03457879
Applications: Debenzylation, C-N and C-O cleavage, alkene hydrogenation
Formula:
Pd Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...