Refractory Metals, plasma standard solution, Specpure®, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
44270-AE
44270-AP
CAAA44270-AEEA
265.44
CAD
CAAA44270-AE
CAAA44270-AP
Refractory Metals, plasma standard solution, Specpure®, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Thermo Scientific
Standards
Cationic Standards
Multi-Element Standards
Molecular Formula: Matrix: 5% HCl/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00151265
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...