Semi Metals, Plasma Standard Solution, Specpure®, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

44268-AE 44268-AP
CAAA44268-AEEA 265.44 CAD
CAAA44268-AE CAAA44268-AP
Semi Metals, Plasma Standard Solution, Specpure®, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Thermo Scientific
Standards Cationic Standards Multi-Element Standards

Molecular Formula: Matrix: 20% HCl/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00145276

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR