Palladium(II) nitrate 1% in nitric acid 10%, Specpure® matrix modifier solution

Supplier: Thermo Scientific Chemicals
Danger

044241-AE 44241-AE
CAAA44241-AEEA 722.06 CAD
CAAA44241-AE AA44241-AE
Palladium(II) nitrate 1% in nitric acid 10%, Specpure® matrix modifier solution
Palladium (II) nitrate
Formula: N₂O₆Pd
MW: 230.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR