Indium gallium alloy (60:40 w%) ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
44240-03
44240-09
CAAA44240-03EA
65.41
CAD
CAAA44240-03
CAAA44240-09
CAAA44240-18
Indium gallium alloy (60:40 w%) ≥99.99% (metals basis)
Indium Gallium alloy (60:40 w%)
Purity: 99.99%
Molecular Formula: Ga:In; 40:60 wt%
Physical Form: Solid
Odor: Odorless
MDL No.: MFCD00144387
Storage Temperature:
Ambient |
MDL Number:
MFCD00144387 EINECS: 231-163-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...