Bismuth lead alloy (55.5:44.5 w%), eutectic, wire, Ø 0.79 mm (0.031 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44238-P2
44238-Q4
CAAA44238-P2EA
1534.18
CAD
CAAA44238-P2
CAAA44238-Q4
Bismuth lead alloy (55.5:44.5 w%), eutectic, wire, Ø 0.79 mm (0.031 in)
Bismuth Lead alloy (55.5:44.5 w%), eutectic
Molecular Formula: Bi:Pb; 55.5:44.5 wt%
Melting Point: 124°
MDL No.: MFCD00801141
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...