Zinc bromide hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

44230-22 44230-36
CAAA44230-22EA 126.87 CAD
CAAA44230-22 CAAA44230-36
Zinc bromide hydrate ≥99.9% (metals basis)
Zinc bromide hydrate

Purity: 99.9% Molecular Formula: ZnBr2.xH2O Formula Weight: 225.19(anhy) Physical Form: Crystalline Merck Reference: 14,10129 UN No.: 3260 MDL No.: MFCD00011294

Formula: Br₂Zn·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 14,10129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR