Platinum 5% on alumina, powder, reduced

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

44222-14 44222-22
CAAA44222-14EA 731.54 CAD
CAAA44222-14 CAAA44222-22
Platinum 5% on alumina, powder, reduced
Platinum

Low surface area


CAS No.: 7440-06-4 MDL No.: MFCD03458043

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR