Poly(vinyl formal)
Supplier: Thermo Scientific Chemicals
Synonyms:
Polyvinylformal, Formvar
44190-14
44190-22
CAAA44190-14EA
132.56
CAD
CAAA44190-14
CAAA44190-22
Poly(vinyl formal)
Poly(vinyl formal)
M.W. 70,000
Soluble in acetone, chloroform, dioxane, THF
CAS No.: 9003-33-2
Physical Form: Granular/ powder
Density: 1.23
Solubility: Soluble in acetone, chloroform, dioxane, THF
MDL No.: MFCD00084463
Formula:
(C₃H₆O₂)n Density: 1,23 Storage Temperature: Ambient |
MDL Number:
MFCD00084463 CAS Number: 9003-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...