2-Methyl-4-trimethylsilyl-1-buten-3-yne ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44189-04
44189-18
CAAA44189-04EA
70.63
CAD
CAAA44189-04
CAAA44189-18
2-Methyl-4-trimethylsilyl-1-buten-3-yne ≥97%
2-Methyl-4-trimethylsilyl-1-buten-3-yne
Purity: 97%
CAS No.: 18387-60-5
Molecular Formula: (CH3)3SiC≡CC(CH3)=CH2
Formula Weight: 138.28
Physical Form: Liquid
Density: 0.778
Boiling Point: 32°/15mm
UN No.: 1993
MDL No.: MFCD00190206
Formula:
(CH₃)₃SiC ΞCC(CH₃)=CH₂ Boiling Pt: 32 °C (15 mmHg) Density: 0,778 Storage Temperature: Ambient |
MDL Number:
MFCD00190206 CAS Number: 18387-60-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...