Bis(1,5-cyclooctadiene)nickel(0) ≥96%

Supplier: Thermo Scientific Chemicals
Danger

44157-04 44157-09
CAAA44157-04EA 170.64 CAD
CAAA44157-04 CAAA44157-09
Bis(1,5-cyclooctadiene)nickel(0) ≥96%
Bis(1,5-cyclooctadiene)nickel (0)

Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene


Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene


Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene

Formula: C₁₆H₂₄Ni
Melting Pt: 60 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00058902
CAS Number: 1295-35-8
EINECS: 215-072-0
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR