Aluminum oxalate hydrate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

44149-09 44149-18 44149-30 44149-A1
CAAA44149-09EA 54.19 CAD
CAAA44149-09 CAAA44149-18 CAAA44149-30 CAAA44149-A1
Aluminum oxalate hydrate ≥99%
Aluminum oxalate hydrate

Purity: 99% CAS No.: 814-87-9 Molecular Formula: C6Al2O12.xH2O Formula Weight: 318.02(anhy) Physical Form: Powder/chunks Appearance: White MDL No.: MFCD00050791

Formula: C₆Al₂O₁₂·xH₂O
MW: 318.017 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00050791
CAS Number: 814-87-9
EINECS: 212-407-2
Merck Index: 14,355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR