Lithium dimethylamide 96
Supplier: Thermo Scientific Chemicals
|
Danger
|
44145-06
44145-14
CAAA44145-06EA
180.12
CAD
CAAA44145-06
CAAA44145-14
Lithium dimethylamide 96
Lithium dimethylamide
Purity: 96%
CAS No.: 3585-33-9
Molecular Formula: C2H6LiN
Formula Weight: 51.02
Physical Form: Powder/Lump
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odor: Amine-like
Melting Point: dec.
Beilstein Reference: 3618233
UN No.: 3393
MDL No.: MFCD00008295
Formula:
C₂H₆LiN MW: 51.017 g/mol Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00008295 CAS Number: 3585-33-9 EINECS: 222-714-3 UN: 3393 ADR: 4.2 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...