Palladium 5% on activated carbon powder standard wetted_50%_Water catalyst_acidic, reduced
Supplier: Thermo Scientific Chemicals
44142-09
44142-18
44142-30
CAAA44142-09EA
260.7
CAD
CAAA44142-09
CAAA44142-18
CAAA44142-30
Palladium 5% on activated carbon powder standard wetted_50%_Water catalyst_acidic, reduced
Palladium
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
Physical Form: 900-1100 m²/g
Odor: Odourless
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03457879 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...