Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Supplier: Thermo Scientific Chemicals

44123-02 44123-04
CAAA44123-02EA 306.52 CAD
CAAA44123-02 CAAA44123-04
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Redox indicator, oxygen sensor


CAS No.: 36309-88-3 Molecular Formula: C72H48Cl2N6Ru Formula Weight: 1169.20 Physical Form: Powder MDL No.: MFCD03095387


Applications: Redox indicator, oxygen sensor

Formula: C₇₂H₄₈Cl₂N₆Ru
MW: 1169.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095387
CAS Number: 36309-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR