3,4,9,10-Perylenetetracarboxylic diimide

Supplier: Thermo Scientific Chemicals

44098-06 44098-14
CAAA44098-06EA 58.62 CAD
CAAA44098-06 CAAA44098-14
3,4,9,10-Perylenetetracarboxylic diimide
3,4,9,10-Perylenetetracarboxylic diimide

CAS No.: 81-33-4 Molecular Formula: C24H10N2O4 Formula Weight: 390.35 Physical Form: Powder MDL No.: MFCD00024144

Formula: C₂₄H₁₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00024144
CAS Number: 81-33-4
EINECS: 201-344-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR